Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GPX8 Recombinant Rabbit Monoclonal Antibody (HL2439)
GREENER_CHOICE

Catalog No. FEMA550532
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEMA550532 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEMA550532 Supplier Invitrogen™ Supplier No. MA550532
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

GPX8 Recombinant Monoclonal Antibody for Western Blot

GPX8 (Probable glutathione peroxidase 8) is also named as GSHPx-8 and belongs to the glutathione peroxidase family. Glutathione peroxidase (GPx) reduces hydroperoxides, including hydrogen peroxides, in the presence of reduced glutathione as a means of protecting organisms from oxidative damage. Several GPx isozymes have been identified in animal cells and these have been classified into different groups according to their cellular location and substrate specificity. In humans, eight types of GPx have been identified from GPx1 to GPx8. The full length GPX8 has 209 amino acids and the molecular weight is 24 kDa.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GPX8
Applications Western Blot
Classification Recombinant Monoclonal
Clone HL2439
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene GPX8
Gene Accession No. Q8TED1
Gene Alias 2310016C16Rik; AU017063; EPLA847; glutathione peroxidase 8; glutathione peroxidase 8 (putative); GPX8; GPx-8; GSHPx-8; probable glutathione peroxidase 8; RGD1307506; UNQ847; UNQ847/PRO1785
Gene Symbols GPX8
Host Species Rabbit
Immunogen Recombinant fragment of human GPX8.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 493869
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.