Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gr-1/Ly-6G Antibody (1A8), DyLight 488, Novus Biologicals™
SDP

Catalog No. NB014962 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB014962 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB014962 Supplier Novus Biologicals Supplier No. NBP281053G
Only null left
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Gr-1/Ly-6G Monoclonal specifically detects Gr-1/Ly-6G in Mouse samples. It is validated for Flow Cytometry.

Specifications

Antigen Gr-1/Ly-6G
Applications Flow Cytometry
Classification Monoclonal
Clone 1A8
Conjugate DyLight 488
Dilution Flow Cytometry
Formulation 50mM Sodium Borate
Gene Alias C6orf23, G6D, LY6-D, LY6G6D, lymphocyte antigen 6 family member G6D, MEGT1, NG25
Gene Symbols LY6G6D
Host Species Rabbit
Immunogen LEW/N rats immunized with 1.5x10E8 Ly-6G-transfected EL4J cells
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Innate Immunity
Primary or Secondary Primary
Gene ID (Entrez) 58530
Test Specificity Recognises the murine GPI-anchored protein Ly6G, a marker of granulocyte differentiation, maturation and, together with Ly6C, a component of the Granulocyte Receptor-1 Antigen (Gr-1). Unlike another commonly used clone, RB6-8C5, 1A8 does not bind Ly6C.
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.