Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Gr-1/Ly-6G Antibody (RB6-8C5), mFluor Violet 500 SE, Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals FAB1037MFV500
Description
Gr-1/Ly-6G Monoclonal antibody specifically detects Gr-1/Ly-6G in Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunocytochemistry, CyTOFSpecifications
Gr-1/Ly-6G | |
Monoclonal | |
mFluor Violet 500 SE | |
50mM Sodium Borate | |
Rat | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
58530 | |
Store at 4C in the dark. | |
IgG2b |
Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunocytochemistry, CyTOF | |
RB6-8C5 | |
Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunocytochemistry, CyTOF-ready | |
C6orf23, G6D, LY6-D, LY6G6D, lymphocyte antigen 6 family member G6D, MEGT1, NG25 | |
The immunogen for this antibody is Ly-6G (Gr-1, Gr1). | |
0.1 mL | |
Primary | |
Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction