Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Gr-1/Ly-6G, Rabbit anti-Mouse, Clone: 1A8, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP2810530.2MG
Description
Gr-1/Ly-6G Monoclonal specifically detects Gr-1/Ly-6G in Mouse samples. It is validated for Flow Cytometry.Specifications
Gr-1/Ly-6G | |
Monoclonal | |
1 mg/mL | |
Flow Cytometry | |
C6orf23, G6D, LY6-D, LY6G6D, lymphocyte antigen 6 family member G6D, MEGT1, NG25 | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Recognises the murine GPI-anchored protein Ly6G, a marker of granulocyte differentiation, maturation and, together with Ly6C, a component of the Granulocyte Receptor-1 Antigen (Gr-1). Unlike another commonly used clone, RB6-8C5, 1A8 does not bind Ly6C. | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
IgG κ |
Flow Cytometry | |
1A8 | |
Unconjugated | |
PBS | |
LY6G6D | |
LEW/N rats immunized with 1.5x10E8 Ly-6G-transfected EL4J cells | |
0.2 mg | |
Innate Immunity | |
58530 | |
Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction