Learn More
GR Rabbit anti-Human, Polyclonal, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A303491A

Description
The recommended shelf life for this product is 1 year from date of receipt. Application Note: For IHC, epitope retrieval with citrate buffer pH 6.0 is recommended for FFPE tissue sections. Based on 100% sequence identity, this antibody is predicted to react with Sheep
Steroid receptors are ligand-dependent, intracellular proteins that stimulate transcription of specific genes by binding to specific DNA sequences following activation by the appropriate hormone. Glucocorticoids are a family of steroids necessary for the regulation of energy metabolism and the immune and inflammatory responses. These compounds exert their effect through their interaction with the glucocoticoid receptor (GR) and that complex′s subsequent association with DNA. All normal mammalian tissues examined to date have been shown to contain glucocorticoid receptor.Specifications
GR | |
Polyclonal | |
Unconjugated | |
NR3C1 | |
GR, GCCR, GCR, Glucocorticoid receptor lymphocyte, GRL, Grl1, NR3C1, Nuclear receptor subfamily 3 group C member 1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
2908 | |
4° C | |
Liquid |
ChIP sequencing (ChIP-seq), Immunohistochemistry, Immunoprecipitation, Western Blot | |
0.2 mg/ml | |
TBS with 0.1% BSA and 0.09% sodium azide | |
P04150 | |
NR3C1 | |
Between 150 and 200 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.