Learn More
GR repeat Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
$518.60
Specifications
Antigen | GR repeat |
---|---|
Concentration | 0.24 mg/mL |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-553-968
![]() ![]() |
Proteintech Group Inc
239781AP150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
This antibody is used to detect the GR repeated sequence. The recombit proteins GST-SNRPD1 and 6xHis tag- SNRPD1 which contains the sequence 'GRGRGRGRGRGRGRGRGR' were used to detect this antibody. Both proteins can be recognized by the antibod
The C9orf72 "GGGGCC" repeat sequence codes five repeat peptide "GA repeat; GAGAGAGAGA", "GP repeat; GPGPGPGPG", "GR repeat; GRGRGRGRG", "AP repeat; APAPAPAPA" and "PR repeat; PRPRPRPRP". It was described previously that aggregated forms of poly-GA and poly-GP proteins do not enter the separation gel.Specifications
GR repeat | |
Western Blot | |
Unconjugated | |
Rabbit | |
Human | |
C9RAN, GGGCC, GR repeat, GRGRGRGRG, GRGRGRGRGR, Poly-GR | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
0.24 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Peptide | |
Primary | |
-20°C |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.