Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NewPath Learning™ Grades 6 - 10 Algebra Skills Curriculum Mastery Flip Chart Set
SCI_ED

Catalog No. S24120 Shop All NewPath Learning Products
Change view
Click to view available options
Grade:
6 to 10
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S24120 6 to 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S24120 Supplier NewPath Learning™ Supplier No. 336002
Add to Cart
Add to Cart

Includes

  • Introduction to Algebra
  • Properties of Addition & Multiplication
  • Introduction to Integers
  • Adding & Subtracting Integers
  • Multiplying & Dividing Integers
  • Solving Equations
  • Multistep Equations
  • Inequalities
  • Introduction to Functions
  • Systems of Equations

Activity Guide featuring questions and exercises.

  • Flip chart mounted on sturdy easel
  • Features 10 double-sided laminated 12 x 18 in. charts
  • Side 1 displays colorful, graphic overview of topic
  • Side 2 features Write-On/Wipe-Off activity Chart features black-line copy-masters, questions, labeling exercises, vocabulary review, etc

Specifications

Grade Level Intermediate - Middle School, High School
Grade 6 to 10
Format Chart
Product Type Algebra Skills Curriculum Mastery Flip Chart Set
Includes
1. Introduction to Algebra
2. Properties of Addition & Multiplication
3. Introduction to Integers
4. Adding & Subtracting Integers
5. Multiplying & Dividing Integers
6. Solving Equations
7. Multistep Equations
8. Inequalities
9. Introduction to Functions
10. Systems of Equations
Certifications Academic standard: California, Florida, National, New York, Texas
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.