Description
- Gradient flow is controlled by a PTFE-coated valve centered between the two chambers
- The outflow channel is fitted with a male luer adapter which can be used with any needle or luer valve
Specifications
Specifications
| For Use With (Application) | Electrophoresis |
| Type | Gradient Marker |
| Volume (English) | 33.8 oz. |
| Volume (Metric) | 1000 mL |