Learn More
Synthware™ Graduated Borosilicate Glass Syringes
GREENER_CHOICESDP
Catalog No. 31502320 Shop All Kemtech Products
Change view
Click to view available options
Volume (Metric):
1 mL
2 mL
10 mL
20 mL
30 mL
50 mL
100 mL
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
31-502-320 10 mL
31-502-317 1 mL
31-502-318 2 mL
31-502-321 20 mL
31-502-322 30 mL
31-502-323 50 mL
31-502-324 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 31-502-320 Supplier Synthware™ Supplier No. S371204
Add to Cart
Add to Cart
Graduated Borosilicate Glass Sample Syringes are constructed of borosilicate glass, and feature permanent graduations with a luer slip tip.
  • Borosilicate glass sample syringes.
  • Permanent graduations.
  • Luer slip tip.
Specifications
Material Borosilicate Glass (Barrel)
For Use With (Application) Precise Sampling
Latex Free Yes
Product Type Syringe
Disposable Reusable
Volume (Metric) 10 mL
For Use With (Equipment) Removable Needle Syringe
Connection Type Leur Slip
Graduated Yes
Sterility Non-sterile