Learn More
Synthware™ Graduated Borosilicate Glass Syringes
GREENER_CHOICESDP
Catalog No. 31502322 Shop All Kemtech Products
Change view
Click to view available options
Volume (Metric):
1 mL
2 mL
10 mL
20 mL
30 mL
50 mL
100 mL
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
31-502-322 30 mL
31-502-317 1 mL
31-502-318 2 mL
31-502-320 10 mL
31-502-321 20 mL
31-502-323 50 mL
31-502-324 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 31-502-322 Supplier Synthware™ Supplier No. S371206
Add to Cart
Add to Cart
Graduated Borosilicate Glass Sample Syringes are constructed of borosilicate glass, and feature permanent graduations with a luer slip tip.
  • Borosilicate glass sample syringes.
  • Permanent graduations.
  • Luer slip tip.
Specifications
Material Borosilicate Glass (Barrel)
For Use With (Application) Precise Sampling
Latex Free Yes
Product Type Syringe
Disposable Reusable
Packaging 1 Ea.
Volume (Metric) 30 mL
For Use With (Equipment) Removable Needle Syringe
Connection Type Leur Slip
Graduated Yes
Sterility Non-sterile