Learn More
Graduated Cylinders
SCI_ED
Catalog No. S95597 Shop All Si Metric Manufacturing Limited Products
Catalog No. S95597 Supplier Si Metric Manufacturing Limited Supplier No. 78120
With embossed calibrations for easy measuring.
  • Durable, transparent polypropylene
  • Wide, stable bases
  • Chemical and acid resistant
  • Autoclavable (dry heat up to 120°C [248°F], wet heat up to 80°C [176°F])
  • Microwavable
Order Info
Available individually or as seven-piece set.
Specifications
Autoclavable Autoclavable
Base Type Wide
Product Type Graduated Cylinder
Max. Temperature (Metric) 120°C