Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GRAF Antibody, Alexa Fluor™ 750, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP232229AF750
Description
GRAF Polyclonal antibody specifically detects GRAF in Human samples. It is validated for Western BlotSpecifications
GRAF | |
Polyclonal | |
Western Blot | |
FLJ42530, GRAFGTPase regulator associated with focal adhesion kinase pp125(FAK), KIAA0621rho GTPase-activating protein 26, Oligophrenin-1-like protein, OPHN1L1, OPHN1LGTPase regulator associated with focal adhesion kinase, Rho GTPase activating protein 26, Rho-type GTPase-activating protein 26 | |
The immunogen this antibody was made to, maps to a region between residue 611 to 661 of human GTPase Regulator Associated with Focal Adhesion Kinase pp125(FAK) using the numbering given in entry NP_055886.1 (GeneID 23092). | |
0.1 mL | |
Neuroscience, Stem Cell Markers | |
23092 | |
Store at 4°C in the dark. | |
IgG |
Western Blot | |
Alexa Fluor 750 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction