Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ GRAF Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA583260
Description
Immunogen sequence: HLSRKKSSDS KPPSCSERPL TLFHTVQSTE KQEQRNSIIN SSLESVSSNP NSILNSSSSL QPNMNSSDPD LAVVKPTRPN SLPPNPSPTS.
Interaction of a cell with the extracellular matrix triggers integrin cell surface receptors to begin signaling cascades that regulate the organization of the actin-cytoskeleton. One of the proteins involved in these cascades is focal adhesion kinase. The protein encoded by this gene is a GTPase activating protein that binds to focal adhesion kinase and mediates the activity of the GTP binding proteins RhoA and Cdc42. Defects in this gene are a cause of juvenile myelomonocytic leukemia (JMML). Multiple transcript variants encoding different isoforms have been found for this gene.
Specifications
GRAF | |
Polyclonal | |
Unconjugated | |
ARHGAP26 | |
1810044B20Rik; 2610010G17Rik; 4933432P15Rik; AI853435; Arhgap26; FLJ42530; GRAF; GRAF1; GTPase regulator associated with focal adhesion kinase; GTPase regulator associated with focal adhesion kinase pp125(FAK); Kiaa0621; mKIAA0621; oligophrenin-1-like protein; OPHN1L; OPHN1L1; Rho GTPase activating protein 26; rho GTPase-activating protein 26; Rho-type GTPase-activating protein 26 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
23092 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9UNA1 | |
ARHGAP26 | |
Recombinant protein corresponding to Human ARHGAP26. Recombinant protein control fragment (Product #RP-95487). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction