Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ GRAF Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA528703
Description
Recommended positive controls: H1299, NIH-3T3, BCL-1. Predicted reactivity: Mouse (100%), Xenopus laevis (100%), Chicken (100%), Bovine (100%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Interaction of a cell with the extracellular matrix triggers integrin cell surface receptors to begin signaling cascades that regulate the organization of the actin-cytoskeleton. One of the proteins involved in these cascades is focal adhesion kinase. The protein encoded by this gene is a GTPase activating protein that binds to focal adhesion kinase and mediates the activity of the GTP binding proteins RhoA and Cdc42. Defects in this gene are a cause of juvenile myelomonocytic leukemia (JMML). Multiple transcript variants encoding different isoforms have been found for this gene.
Specifications
GRAF | |
Polyclonal | |
Unconjugated | |
ARHGAP26 | |
1810044B20Rik; 2610010G17Rik; 4933432P15Rik; AI853435; Arhgap26; FLJ42530; GRAF; GRAF1; GTPase regulator associated with focal adhesion kinase; GTPase regulator associated with focal adhesion kinase pp125(FAK); Kiaa0621; mKIAA0621; oligophrenin-1-like protein; OPHN1L; OPHN1L1; Rho GTPase activating protein 26; rho GTPase-activating protein 26; Rho-type GTPase-activating protein 26 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
23092, 71302 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
1 mg/mL | |
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 | |
Q6ZQ82, Q9UNA1 | |
ARHGAP26 | |
Synthetic peptide corresponding to a region within amino acids 752 and 814 of Human GRAF. | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction