Learn More
GRAF1 Rabbit anti-Human, Polyclonal, Bioss
SDP
Rabbit Polyclonal Antibody
Supplier:  Bioss BS11248R
Encompass_Preferred
Catalog No. 50-198-1205
Only null left
Add to Cart
Description
Description
Interaction of a cell with the extracellular matrix triggers integrin cell surface receptors to begin signaling cascades that regulate the organization of the actin-cytoskeleton. One of the proteins involved in these cascades is focal adhesion kinase. The protein encoded by this gene is a GTPase activating protein that binds to focal adhesion kinase and mediates the activity of the GTP binding proteins RhoA and Cdc42. Defects in this gene are a cause of juvenile myelomonocytic leukemia (JMML). Multiple transcript variants encoding different isoforms have been found for this gene.
Specifications
Specifications
GRAF1
Polyclonal
Unconjugated
ARHGAP26
1810044B20Rik; 2610010G17Rik; 4933432P15Rik; AI853435; ARHGAP26; FLJ42530; GRAF; GRAF1; GTPase regulator associated with focal adhesion kinase; GTPase regulator associated with focal adhesion kinase pp125(FAK); KIAA0621; mKIAA0621; Oligophrenin-1-like protein; OPHN1L; OPHN1L1; Rho GTPase activating protein 26; Rho GTPase-activating protein 26; Rho-type GTPase-activating protein 26
Rabbit
Protein A
RUO
23092
-20°C
Liquid
Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
1 μg/mL
PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Q9UNA1
ARHGAP26
KLH conjugated synthetic peptide derived from human GRAF1.
100 μL
Primary
Human
Antibody
IgG
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.