Learn More
United Scientific Supplies Graham Condensers, 24/40 Joint
SDPSCI_ED
Designed for use in distillation applications
Supplier:  United Scientific Supplies CNG400
 View more versions of this product
Catalog No. S43786
Only null left
Add to Cart
Description
Description
  • Designed for use in distillation applications
  • Constructed of borosilicate glass
  • A coiled inner tube provides additional surface area for highly efficient cooling
  • Socket is 24/40 with a screw thread connector
Specifications
Specifications
Screw Thread
Graham Condenser
Distillation Glassware
400 mm
Borosilicate Glass
Distillation
Spiral
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.