Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GRAIL/RNF128 Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody has been used in 3 publications

$206.50 - $499.50

Specifications

Antigen GRAIL/RNF128
Concentration 1.0 mg/mL
Dilution Western Blot 0.5-2 ug/ml, Immunohistochemistry, Immunocytochemistry/Immunofluorescence reported in scientific literature (PMID 24356810), Immunohistochemistry-Paraffin 5-10 ug/ml
Applications Western Blot, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP224610SS
SDP
View Documents
Novus Biologicals
NBP224610SS
0.025 mg
Each for $206.50
Only null left
Add to Cart
 
NBP224610
SDP
View Documents
Novus Biologicals
NBP224610
0.1 mg
Each for $499.50
Only null left
Add to Cart
 
Description

Description

GRAIL/RNF128 Polyclonal specifically detects GRAIL/RNF128 in Human, Mouse, Rat, Canine, Equine, Primate samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications

Specifications

GRAIL/RNF128
Western Blot 0.5-2 ug/ml, Immunohistochemistry, Immunocytochemistry/Immunofluorescence reported in scientific literature (PMID 24356810), Immunohistochemistry-Paraffin 5-10 ug/ml
Polyclonal
Purified
RUO
PBS with 0.05% Sodium Azide
EC 6.3.2, EC 6.3.2.-, FLJ23516, Gene related to anergy in lymphocytes protein, ring finger protein 128GRAILE3 ubiquitin-protein ligase RNF128
RNF128
IgG
Protein A purified
The amino acid sequence used as immunogen is 100% homologous in human, horse, chimpanzee, monkey and orangutan, 94% homologous in cow, 88% homologous in cat and 83% homologous in opossum, rat and mouse.
1.0 mg/mL
Western Blot, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Unconjugated
Rabbit
Human, Mouse, Rat, Canine, Equine, Primate
Q8TEB7
79589
A synthetic peptide corresponding to amino acids portion of amino acids 227-269 of human GRAIL was used as immunogen, GenBank no. NP_919445.1.
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.