Learn More
MilliporeSigma™ Gram-Color modified
Shop All MilliporeSigma Products
Click to view available options
Quantity:
1 Set
Description
- Phenol-free
- Gram-positve microorganisms: Dark blue
- Gram-negative microorganisms:Red
Specifications
Specifications
Quantity | 1 Set |
Packaging Type | Fibre Case |