Learn More
Invitrogen™ Gram Positive Bacteria LTA Monoclonal Antibody (3804)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA547521
Description
Specificity: Reactive with lipoteichoic acid (LTA) of many gram positive bacteria: Listeria monocytogenes (all serotypes), Streptococcus pneumoniae, Staphylococcus aureus, Staphylococcus epidermidis, Enterococcus faecalis, Bacillus cereus, Bacillus subtilis, and group A Strep. Purified preparations consist of >90% pure mouse monoclonal antibody which has been purified from ascites fluid by protein A chromatography. The final preparation is formulated to a protein concentration of 100 μg/mL in 0.01 M phosphate buffered saline, pH 7.2 and contains 0.1% sodium azide. Each vial contains 1.0 mL. This product contains no stabilizing proteins and should be stored at 4C until ready for use. Working dilution must be determined by the user. Suggested starting ranges are 1:10-1:50 for ICC/IF and 1:20-1:200. for ELISA.

Specifications
Gram Positive Bacteria LTA | |
Monoclonal | |
100 μg/mL | |
PBS with 0.1% sodium azide; pH 7.2 | |
Mouse | |
Protein A | |
RUO | |
Bacteria | |
Antibody | |
IgG1 |
ELISA, Immunocytochemistry | |
3804 | |
Unconjugated | |
Lipoteichoic acid | |
Proprietary sequence. | |
100 μg | |
Primary | |
4°C | |
Liquid |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.