Learn More
Invitrogen™ Gram Positive Bacteria LTA Monoclonal Antibody (G35C)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA17401
Description
The MA1-7401 antibody reacts with lipoteichoic acid from many gram positive bacteria. Reactive with Listeria monocytogenes (all serotypes), Streptococcus pneumoniae, Staphylococcus aureus, Staphylococcus epidermidis, Enterococcus faecium, Bacillus cereus, Bacillus subtilis, group B Streptococcus (weak). Non-reactive with Clostridium perfringens. Other species not tested. MA1-7401 has been successfully used in ELISA and Immunofluorescence applications. Suggested but not tested for Western blot and immunohistochemistry applications.

Specifications
Gram Positive Bacteria LTA | |
Monoclonal | |
0.1 mg/mL | |
PBS with 0.1% sodium azide; pH 7.2 | |
Mouse | |
Protein A | |
RUO | |
Bacteria | |
Antibody | |
IgG1 |
ELISA, Immunocytochemistry | |
G35C | |
Unconjugated | |
Lipoteichoic acid | |
Lipoteichoic acid consistent with that of gram positive bacteria. | |
100 μg | |
Primary | |
4°C | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.