Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Innovating Science™ Gram Stain Kit
SCI_ED

Catalog No. s37740 Shop All Aldon Products
Click to view available options
Quantity:
1
WARNING: This product can expose you to chemicals including Crystal Violet and Methanol/ Methyl Isobutyl Ketone, which are known to the State of California to cause cancer and reproductive harm.
WARNING: This product can expose you to chemicals including Crystal Violet and Methanol/ Methyl Isobutyl Ketone, which are known to the State of California to cause cancer and reproductive harm.

Includes

1 X 500mL Dilute Lugols, 1 X 500mL Crystal Violet, 1 X 500mL Safranin O 1% Aqueous, 2L Ethanol 95%

To determine if bacteria are Gram-positive or Gram-negative.

This kit is used to determine if bacteria are Gram-positive or Gram-negative.

Specifications

Type Gram Stain Kit
Quantity 1
Includes 1 X 500mL Dilute Lugols, 1 X 500mL Crystal Violet, 1 X 500mL Safranin O 1% Aqueous, 2L Ethanol 95%
For Use With (Application) To determine if bacteria are Gram-positive or Gram-negative.
Storage Requirements Ambient
Volume (Metric) 500 mL Dilute Lugols, 500 mL Crystal Violet, 500 mL Safranin O 1% Aqueous, 2 x 1 L Ethanol 95%
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.