Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GRAMD1A Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP232148
Description
GRAMD1A Polyclonal specifically detects GRAMD1A in Human samples. It is validated for Western Blot, Immunoprecipitation.Specifications
GRAMD1A | |
Polyclonal | |
Unconjugated | |
Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide | |
FLJ22411, FLJ90346, GRAM domain containing 1A, GRAM domain-containing protein 1A, KIAA1533 | |
Rabbit | |
Affinity Purified | |
RUO | |
57655 | |
Store at 4C. Do not freeze. |
Western Blot, Immunoprecipitation | |
1.0 mg/mL | |
Western Blot 1:1000 - 1:5000, Immunoprecipitation 2 - 10 ug/mg lysate | |
NP_065946.2 | |
GRAMD1A | |
The immunogen this antibody was made to, maps to a region between residue 674 to 724 of human GRAM Domain Containing 1A using the numbering given in entry NP_065946.2 (GeneID 57655). | |
100 μL | |
Primary | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction