Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GRAMD1C Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA552928
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA552928 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA552928 Supplier Invitrogen™ Supplier No. PA552928
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: VSTDLKYRKQ PWGLVKSLIE KNSWSSLEDY FKQLESDLLI EESVLNQAIE DPGKLTGLRR RRRTFNRTAE TVPKLSSQHS SGDVGLGAKG Highest antigen sequence identity to the following orthologs: Mouse - 80%, Rat - 79%.

TRUSTED_SUSTAINABILITY

Specifications

Antigen GRAMD1C
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene GRAMD1C
Gene Accession No. Q8IYS0
Gene Alias GRAM domain containing 1C; GRAM domain-containing protein 1C; GRAMD1C; Protein Aster-C; UNQ2543/PRO6095
Gene Symbols GRAMD1C
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human GRAMD1C. Recombinant protein control fragment (Product #RP-90496).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54762
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.