Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ GRAMD3 Polyclonal Antibody

Goat Polyclonal Antibody
Supplier: Invitrogen™ PA519380
Description
This antibody is predicted to react with bovine, canine and rat based on sequence homology. This antibody is tested in Peptide ELISA: antibody detection limit dilution 4,000.
Specifications
| GRAMD3 | |
| Polyclonal | |
| Unconjugated | |
| GRAMD2B | |
| 9030613F08Rik; 9130427A09Rik; AW061306; FLJ21313; GRAM domain containing 3; GRAM domain-containing protein 2B; GRAM domain-containing protein 3; GRAMD2B; GRAMD3; HCV NS3-transactivated protein 2; hepatitis C virus nonstructural protein 3-transactivating protein 2; NS3TP2 | |
| Goat | |
| Ammonium sulfate precipitation | |
| RUO | |
| 107022, 65983 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
| Q6PEM6, Q96HH9 | |
| GRAMD2B, Gramd3 | |
| Synthetic peptide sequence (QTPESENSRD) corresponding to the internal amino acids of GRAMD3 (aa 274-283). | |
| 100 μg | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction