Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GRAMD4 Antibody (PCRP-GRAMD4-1A10), Alexa Fluor™ 750, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP314282AF750
This item is not returnable.
View return policy
Description
GRAMD4 Monoclonal antibody specifically detects GRAMD4 in Human samples. It is validated for Flow Cytometry, ImmunofluorescenceSpecifications
GRAMD4 | |
Monoclonal | |
Alexa Fluor 750 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified | |
RUO | |
23151 | |
Store at 4°C in the dark. | |
Purified |
Flow Cytometry, Immunofluorescence | |
PCRP-GRAMD4-1A10 | |
Flow Cytometry, Immunocytochemistry/Immunofluorescence | |
GRAM domain containing 4, GRAM domain-containing protein 4, KIAA0767 | |
Recombinant full-length human GRAMD4 protein (Uniprot: Q6IC98) | |
0.1 mL | |
Primary | |
Human | |
Antibody | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction