Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GRAMD4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA598543
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA598543 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA598543 Supplier Invitrogen™ Supplier No. PA598543
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

GRAMD4 Polyclonal Antibody for Western Blot, IHC (P), ELISA

GRAMD4 also termed as Death inducing protein is a 578 amino-acid protein, which contains 1 GRAM domain. GRAMD4 as a multi-pass membrane protein localizes in the Mitochondrion membrane. Overexpression of GRAMD4 results in a strong apoptotic response involving caspase-3 activation and cleavage of poly (ADP-ribose)-polymerase. GRAMD4 is expressed in lung and in primary lung squamous cell carcinoma (LSCC) and shows up-regulation in mitochondria by E2F-1 after addition of 4-hydroxytamoxifen. This evidence suggests that GRAMD4 may be a potential target for cancer therapies.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GRAMD4
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 5 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene GRAMD4
Gene Accession No. Q6IC98, Q8CB44
Gene Alias 9930016O13; dA59H18.1; death-inducing protein; death-inducing-protein; DIP; dJ439F8.1; GRAM domain containing 4; GRAM domain-containing protein 4; GRAM domain-containing protein 4 (GRAMD4); Gramd4; Kiaa0767
Gene Symbols GRAMD4
Host Species Rabbit
Immunogen Recombinant Human GRAM domain-containing protein 4 protein (23-161AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 223752, 23151
Target Species Human, Mouse
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.