Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GRAMD4 Rabbit anti-Human, Polyclonal, Proteintech
SDP

Catalog No. 501734287 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-4287 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-4287 Supplier Proteintech Group Inc Supplier No. 242991AP150UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

GRAMD4 also termed as Death inducing protein is a 578 amino-acid protein, which contains 1 GRAM domain. GRAMD4 as a multi-pass membrane protein localizes in the Mitochondrion membrane. Overexpression of GRAMD4 results in a strong apoptotic response involving caspase-3 activation and cleavage of poly (ADP-ribose)-polymerase. GRAMD4 is expressed in lung and in primary lung squamous cell carcinoma (LSCC) and shows up-regulation in mitochondria by E2F-1 after addition of 4-hydroxytamoxifen. This evidence suggests that GRAMD4 may be a potential target for cancer therapies.

Specifications

Antigen GRAMD4
Applications Western Blot
Classification Polyclonal
Concentration 0.59 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene GRAMD4
Gene Accession No. Q6IC98
Gene Alias dA59H18.1, Death inducing protein, DIP, GRAM domain containing 4, GRAMD4, KIAA0767
Gene Symbols GRAMD4
Host Species Rabbit
Immunogen GRAMD4 Fusion Protein Ag19140
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23151
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.