Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NewPath Learning™ Grand Canyon 3D Model Making Kit
SCI_ED

Catalog No. S436052 Shop All NewPath Learning Products
Change view
Click to view available options
Grade:
6 to 10
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S436052 6 to 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. S436052 Supplier NewPath Learning™ Supplier No. 247819
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

The Grand Canyon 3D Model Making Kit will fascinate students as they explore the Grand Canyon, its numerous horizontal layers of different rock types, and how it was formed.

  • The Grand Canyon is a vast and spectacular mile-deep slit in the Earth's surface carved over millions of years by the Colorado River!
  • Your students will find the Grand Canyon fascinating as they learn how its numerous horizontal layers of different rock types have been laid down over two billion years.
  • First students make their model using the colorful parts provided.
  • They will then use their model to look at how the Grand Canyon formed and what the different layers represent.
  • Through this process, they will learn that the layers were formed by seas, rivers, dry deserts, and entire mountain ranges.
  • The Model Making Kit includes enough materials for students to assemble up to 5 paper models working cooperatively.
  • The comprehensive Teacher Resource Guide included provides illustrated readings, activities, key vocabulary and an assessment, along with teaching opportunities for students to Engage, Explore, Explain, Extend and Evaluate their understanding of the concepts presented.

Specifications

Grade Level Intermediate - Middle School, High School
Grade 6 to 10
Product Type 3D Model
Includes 5 Models, Teacher Resource Guide, Vinyl Pouch
Width (English) 9 in.
Height (English) 12 in.
Depth (English) 2 in.
Material Paper
Languages English
For Use With (Application) Model Assembly
Subject STEM, Geology
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.