Learn More
Eisco™ Granite Specimen
SCI_ED
Catalog No. S24760 Shop All Eisco Products
Change view
Click to view available options
Quantity:
1 Ea.
10/Pk.
12/Pk.
Specimen Size:
2 to 3 cm
7 to 8 cm
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Specimen Size
S24760 1 Ea. 2 to 3 cm
S26609 10/Pk. 2 to 3 cm
S24761 12/Pk. 2 to 3 cm
S26668 1 Ea. 7 to 8 cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. S24760 Supplier Eisco™ Supplier No. ESNG0045
Add to Cart
Add to Cart
Granite Specimen is a must-have sample when discussing rock types, rock cycle, formation, classification, and identification of igneous rocks.
  • Pink granite sample
  • Large crystals in this felsic plutonic rock are for use in lessons in mineral identification and bulk rock chemistry
  • For comparing cooling rates between samples with differently-sized crystals
  • Correlates well to Next Generation Science Standards Evidence Statements
  • Large crystals are indicative of slow cooling rate, and make mineral identification simple
Specifications
Product Type Igneous Rock
For Use With (Application) Comparing cooling rates between samples with differently-sized crystals
Packaging Clear labeled and barcoded bag
Quantity 1 Ea.
Specimen Size 2 to 3 cm
Specimen Type Granite