Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Granulin Antibody (CL5692), Novus Biologicals™

Mouse Monoclonal Antibody
$382.00 - $610.00
Specifications
Antigen | Granulin |
---|---|
Clone | CL5692 |
Dilution | Immunohistochemistry-Paraffin 1:1000 - 1:2500 |
Classification | Monoclonal |
Conjugate | Unconjugated |
Description
Granulin Monoclonal specifically detects Granulin in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
Granulin | |
Immunohistochemistry-Paraffin 1:1000 - 1:2500 | |
Unconjugated | |
Mouse | |
Human | |
2896 | |
This antibody was developed against a recombinant protein corresponding to amino acids: SCPDGYTCCRLQSGAWGCCPFTQAVCCEDHIHCCPAGFTCDTQKGTCEQGPHQVPWMEKAPAHLSLPDPQALKRDVPCDNVSSCPSSDTCCQLTSGEWGCCPIPEAVCCSDHQHCCPQGYTCVAEGQCQRGSEIVAGL | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
CL5692 | |
Monoclonal | |
Purified | |
Cell Cycle and Replication | |
acrogranin, GEP, GP88, granulin, granulin-epithelin, granulins, PC cell-derived growth factor, PCDGF, PEPI, PGRN, proepithelin, progranulin | |
GRN | |
IgG1 | |
Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title