Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Granulin Antibody (OTI3H6), FITC, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP270847F
Description
Granulin Monoclonal specifically detects Granulin in Human samples. It is validated for Immunohistochemistry.Specifications
Granulin | |
Monoclonal | |
FITC | |
PBS with 0.05% Sodium Azide | |
GRN | |
Recombinant protein fragment corresponding to amino acids 327-593 of human Granulin (NP_002078) produced in E.coli. | |
0.1 mL | |
Primary | |
Human | |
IgG1 |
Immunohistochemistry | |
OTI3H6 | |
Immunohistochemistry | |
acrogranin, GEP, GP88, granulin, granulin-epithelin, granulins, PC cell-derived growth factor, PCDGF, PEPI, PGRN, proepithelin, progranulin | |
Mouse | |
Affinity Purified | |
Cell Cycle and Replication | |
2896.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction