Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Granulin Antibody Pair [HRP]
SDP

Catalog No. p-200083689
Click to view available options
Quantity:
15 Plates
5 Plates

Antibody Pair

Specifications

Assay Range 39.06 to 2500 pg/mL (example only, lot dependent)
Assay Sensitivity 39.06 pg/mL (example only; lot dependent)
Conjugate HRP
Detection Method Sandwich ELISA
Immunoassay Kit Format Sandwich ELISA Kit
Product Type Sandwich ELISA
Specificity This solid phase sandwich ELISA utilizes a monoclonal antibody specific for Mouse Granulin.
Target Species Mouse
For Use With (Application) ELISA, Sandwich ELISA
Gene ID (Entrez) 2896
Gene Symbol GRN
Quantity 15 Plates
Research Discipline Cell Cycle and Replication
Storage Requirements Storage of components varies. See protocol for specific instructions.
Synonym acrogranin, GEP, GP88, granulin, granulin-epithelin, granulins, PC cell-derived growth factor, PCDGF, PEPI, PGRN, proepithelin, progranulin
Target Mouse
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.