Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Granulin Rabbit anti-Human, Clone: 8O5Q5, Novus Biologicals™

Rabbit Monoclonal Antibody
$205.50 - $458.50
Specifications
Antigen | Granulin |
---|---|
Clone | 8O5Q5 |
Applications | Western Blot |
Classification | Monoclonal |
Conjugate | Unconjugated |
Description
Granulin Monoclonal antibody specifically detects Granulin in Human samples. It is validated for Western BlotSpecifications
Granulin | |
Western Blot | |
Unconjugated | |
Rabbit | |
Cell Cycle and Replication | |
PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
2896 | |
IgG | |
Affinity purified |
8O5Q5 | |
Monoclonal | |
Purified | |
RUO | |
Human | |
acrogranin, GEP, GP88, granulin, granulin-epithelin, granulins, PC cell-derived growth factor, PCDGF, PEPI, PGRN, proepithelin, progranulin | |
Recombinant fusion protein containing a sequence corresponding to amino acids 20-200 of human Granulin (NP_002078.1). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title