Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Granulocyte Macrophage-Colony Stimulating Factor Monoclonal Antibody (ANT-129), QED Bioscience™
SDP

Catalog No. 502278121
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
500 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-227-8121 1 mg
50-227-8122 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 50-227-8121 Supplier Leinco Technologies Inc Supplier No. 501721000UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Granulocyte Macrophage-Colony Stimulating Factor Monoclonal antibody specifically detects Granulocyte Macrophage-Colony Stimulating Factor in Human samples. It is validated for ELISA

Specifications

Antigen Granulocyte Macrophage-Colony Stimulating Factor
Applications ELISA
Classification Monoclonal
Clone ANT-129
Concentration 0.5-1.0 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene CSF2
Gene Accession No. P04141
Gene Alias colony stimulating factor 2; colony stimulating factor 2 (granulocyte-macrophage); colony-stimulating factor; CSF; CSF2; Csfgm; cytokine; GMCSF; Gm-CSf; granulocyte macrophage colony stimulating factor; granulocyte macrophage-colony stimulating factor; granulocyte-macrophage colony stimulating factor 2; granulocyte-macrophage colony-stimulating factor; MGC131935; MGC138897; MGC151255; MGC151257; MGI-IGM; M-GM-CSF; molgramostin; put. GM-CSF; sargramostim
Host Species Mouse
Immunogen Recombinant human GM-CSF
Purification Method Protein A
Quantity 1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1437
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.