Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Granulocyte Marker Antibody (SPM250) - IHC-Prediluted, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP248057
Description
Granulocyte Marker Monoclonal specifically detects Granulocyte Marker in Human, Rhesus Macaque samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.Specifications
Granulocyte Marker | |
Monoclonal | |
Unconjugated | |
10 mM PBS with 0.05% BSA with 0.05% Sodium Azide | |
Nuclei from pokeweed mitogen stimulated human peripheral blood lymphocytes. | |
7 mL | |
Primary | |
Human, Primate | |
Purified |
Immunohistochemistry (Paraffin) | |
SPM250 | |
Immunohistochemistry-Paraffin | |
Mouse | |
Protein A or G purified | |
RUO | |
Recognizes an unidentified antigen in the cytoplasm of mature granulocytes. It shows no reactivity with any other cell type in human tissues. Markers of myeloid cells are useful in the identification of different levels of cellular differentiation. monoclonal antibody BM-2 reacts with early precursor and mature forms of human myeloid cells. This monoclonal antibody is useful for the detection of myeloid leukemias and granulocytic sarcomas. It can be used as a marker of granulocytes in normal tissues or inflammatory processes. | |
Store at 4C. | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction