Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Granulocyte Marker Antibody (SPM250), mFluor Violet 500 SE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP234750MFV500
Description
Granulocyte Marker Monoclonal antibody specifically detects Granulocyte Marker in Human, Macaque samples. It is validated for Flow CytometrySpecifications
Granulocyte Marker | |
Monoclonal | |
mFluor Violet 500 SE | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified | |
RUO | |
Human, Macaque | |
Purified |
Flow Cytometry | |
SPM250 | |
Flow Cytometry | |
Granulocytes;PML;PMN | |
Nuclei from pokeweed mitogen stimulated human peripheral blood lymphocytes. | |
0.1 mL | |
Primary | |
Store at 4C in the dark. | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction