Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Granulocytes Monoclonal Antibody (6D10), FITC
GREENER_CHOICE

Catalog No. PIMA516533
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA516533 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA516533 Supplier Invitrogen™ Supplier No. MA516533
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

For FACS analysis, use 10 μL of the suggested working dilution to label 1x10^6 cells in 100 μL. Mouse anti Pig Granulocytes antibody, clone 6D10 recognizes a approximately 60 kDa antigen on porcine granulocytes of the neutrophil lineage, acting as a reliable tool for their analysis and isolation, without contamination from other cells.

Granulocytes are a category of white blood cells characterized by the presence of granules in their cytoplasm. They are also called polymorphonuclear leukocytes (PMN or PML) because of the varying shapes of the nucleus, which is usually lobed into three segments. In common parlance, the term polymorphonuclear leukocyte often refers specifically to neutrophil granulocytes, the most abundant of the granulocytes. Granulocytes or PMN are released from the bone marrow by the regulatory complement proteins.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Granulocytes
Applications Flow Cytometry
Classification Monoclonal
Clone 6D10
Concentration 0.1 mg/mL
Conjugate FITC
Formulation PBS with 1% BSA and 0.09% sodium azide
Gene Alias C17orf33; colony stimulating factor 3; colony stimulating factor 3 (granulocyte); CSF3; CSF3OS; filgrastim; GCSF; G-CSF; Granulocyte colony-stimulating factor; Granulocyte Marker; granulocyte-colony stimulating factor; lenograstim; Pluripoietin; PML; PMN
Host Species Mouse
Immunogen Porcine bone marrow haematopoietic cells (BMHC).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Pig
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. Store in the dark.
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.