Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Granulocytes Mouse Anti-Rat, Brilliant Violet 711, Clone: RP-1, BD Optibuild™

Catalog No. BDB743057 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB743057 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB743057 Supplier BD Biosciences Supplier No. 743057
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The RP-1 antibody reacts with rat peritoneal and peripheral blood neutrophils and bone marrow cells. It does not bind rat macrophages, rat eosinophils, or peritoneal neutrophils from mice, rabbits, guinea pigs, or human peripheral blood as determined by antibody adsorption experiments. Flow cytometric analysis of peripheral blood leukocytes indicates that all granulocytes (cells having high side scatter), but not lymphocytes or monocytes (low side-scatter cells), express that antigen recognized by RP-1 mAb. Expression of the RP-1 antigen on rat peritoneal neutrophils is enhanced by stimulation with PMA or ConA. Immunoprecipitation of rat PMA-activated and non-treated neutrophil membranes with the RP-1 mAb produced two bands of approximately 85 kDa in m.w. However, the RP-1 antigen has not been biochemically characterized.

The antibody was conjugated to BD Horizon™ BV711 which is part of the BD Horizon Brilliant™ Violet family of dyes. This dye is a tandem fluorochrome of BD Horizon BV421 with an Ex Max of 405nm and an acceptor dye with an Em Max at 711nm. BD Horizon BV711 can be excited by the violet laser and detected in a filter used to detect Cy™5.5 / Alexa Fluor™ 700-like dyes (eg, 712/20nm filter). Due to the excitation and emission characteristics of the acceptor dye, there may be moderate spillover into the Alexa Fluor™ 700 and PerCP-Cy5.5 detectors. However, the spillover can be corrected through compensation as with any other dye combination. BV711 is a tandem fluorochrome of BD Horizon BV421 and an acceptor dye with an emission maximum at 711 nm. This dye offers a very bright choice for the violet laser. Due to the excitation and emission characteristics of the acceptor dye, there may be moderate spillover into the Alexa Fluor™ 700 and PerCP-Cy5.5 detectors. BV711 will also have moderate spillover into the BD Horizon Brilliant™ Violet 786 detector.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Granulocytes
Classification Monoclonal
Clone RP-1
Concentration 0.2mg/mL
Conjugate Brilliant Violet 711
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Mouse
Immunogen WKA/Hok rat peritoneal neutrophils
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Mouse
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG2a κ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.