Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Granulysin Antibody (B-R32), DyLight 350, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP318105UV
Description
Granulysin Monoclonal antibody specifically detects Granulysin in Human samples. It is validated for ELISASpecifications
Granulysin | |
Monoclonal | |
DyLight 350 | |
D2S69Elymphocyte-activation gene 2, granulysin, LAG2, LAG-2, Lymphokine LAG-2, NKG5519, Protein NKG5, TLA519T-cell activation protein 519, T-lymphocyte activation gene 519 | |
Recombinant human Granulysin (NKG5 and LAG-2). Hybridoma: Myeloma X63/AG.8653 x Balb/c node cells | |
0.1 mL | |
Adaptive Immunity, Immunology | |
10578 | |
Store at 4°C in the dark. | |
IgG2b κ |
ELISA | |
B-R32 | |
50mM Sodium Borate | |
Mouse | |
Affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction