Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Granulysin Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA563509
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA563509 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA563509 Supplier Invitrogen™ Supplier No. PA563509
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: RLSPEYYDLA RAHLRDEEKS CPCLAQEGPQ GDLLTKTQEL GRDYRTCLTI VQKLKKMVDK P Highest antigen sequence identity to the following orthologs: Mouse - 28%, Rat - 28%.

GNLY is a member of the saposin-like protein (SAPLIP) family and is located in the cytotoxic granules of T cells, which are released upon antigen stimulation. GNLY is present in cytotoxic granules of cytotoxic T lymphocytes and natural killer cells, and it has antimicrobial activity against M. tuberculosis and other organisms.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Granulysin
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene GNLY
Gene Accession No. P22749
Gene Alias D2S69E; GNLY; Granulysin; LAG2; LAG-2; lymphocyte-activation gene 2; Lymphokine; lymphokine LAG-2; Lymphokine NKG5519; NKG5; Protein NKG5; T-cell activation protein 519; TLA519; T-lymphocyte activation gene 519
Gene Symbols GNLY
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human GNLY. Recombinant protein control fragment (Product #RP-103315).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10578
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.