Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Granzyme A Antibody (CB9), Alexa Fluor™ 532, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP314619AF532
This item is not returnable.
View return policy
Description
Granzyme A Monoclonal antibody specifically detects Granzyme A in Human samples. It is validated for Flow Cytometry, ImmunofluorescenceSpecifications
Granzyme A | |
Monoclonal | |
Alexa Fluor 532 | |
50mM Sodium Borate | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Antibody | |
IgG1 κ |
Flow Cytometry, Immunofluorescence | |
CB9 | |
Flow Cytometry, Immunocytochemistry/Immunofluorescence | |
CTL tryptase, CTLA3Granzyme A (Cytotoxic T-lymphocyte-associated serine esterase-3; Hanukah factorserine protease), Cytotoxic T-lymphocyte proteinase 1, EC 3.4.21, fragmentin-1, granzyme A, granzyme A (granzyme 1, cytotoxic T-lymphocyte-associated serine esterase 3), Granzyme-1, H factor, Hanukkah factor, HF, HFSPEC 3.4.21.78 | |
Full length human granzyme A | |
0.1 mL | |
Adaptive Immunity, Apoptosis, Cancer, Immunology, Innate Immunity, Tumor Suppressors | |
3001 | |
Store at 4°C in the dark. | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction