Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Granzyme A Monoclonal Antibody (GzA-3G8.5), PE-Cyanine7, eBioscience™, Invitrogen™

Mouse Monoclonal Antibody

$188.00 - $453.00

Specifications

Antigen Granzyme A
Clone GzA-3G8.5
Concentration 0.2 mg/mL
Applications Flow Cytometry
Classification Monoclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
25-583-180
Encompass_Preferred
View Documents
Invitrogen™
25583180
25 μg
Each for $188.00
Only null left
Add to Cart
 
50-112-3394
Encompass_Preferred
View Documents
Invitrogen™
25583182
100 μg
Each for $453.00
Only null left
Add to Cart
 
Description

Description

Description: This Gza-3G8.5 monoclonal antibody reacts with mouse Granzyme A. Granzyme A is the most abundantly expressed of the ten granzyme serine proteases that have been identified in mice. Granzymes are proteins released from the granules of NK cells and cytotoxic T lymphocytes that induce death in target cells by cleavage of intracellular substrates and play a critical role in immune defense against viruses, tumors, and intracellular bacteria. Granzyme A activates caspase-independent cell death pathways morphologically similar to apoptosis and characterized by mitochondrial and DNA damage. It may also play a role in inflammation, as the precursor form of IL-1 beta (pro-IL-1 beta) is among its target substrates. Granzyme A shares overlapping substrate specificity with the closely-related Granzyme K, which is believed to account for the minimal decrease in cytotoxicity of Granzyme A-deficient CTLs. Applications Reported: This GzA-3G8.5 antibody has been reported for use in intracellular staining followed by flow cytometric analysis. Applications Tested: This GzA-3G8.5 antibody has been tested by intracellular staining followed by flow cytometric analysis of mouse splenocytes. This can be used at less than or equal to 0.06 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test.

Granzyme A Monoclonal antibody specifically detects Granzyme A in Mouse samples. It is validated for Flow Cytometry.
Specifications

Specifications

Granzyme A
0.2 mg/mL
Monoclonal
Liquid
RUO
PBS with 0.09% sodium azide; pH 7.2
Autocrine thymic lymphoma granzyme-like serine protease; AW494114; BLT esterase; CTL tryptase; CTLA3; CTLA-3; Cytotoxic T-lymphocyte proteinase 1; Cytotoxic T-lymphocyte-associated serine esterase-3; fragmentin-1; granzyme 1; granzyme A; Granzyme A (Cytotoxic T-lymphocyte-associated serine esterase-3; Hanukah factor serine protease); granzyme A (granzyme 1, cytotoxic T-lymphocyte-associated serine esterase 3); Granzyme-1; GZMA; H factor; Hanukah factor; Hanukah factor serine protease); Hanukkah factor; Hf; HFSP; Mtsp-1; SE1; serine esterase 1; t cell-specific serine protease 1; TSP1; TSP-1
GZMA
Primary
4°C, store in dark, DO NOT FREEZE!
GZMA
GzA-3G8.5
Flow Cytometry
PE-Cyanine7
Mouse
Mouse
P11032
14938
IgG2b κ
Affinity chromatography
Antibody
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only.

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.