Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Granzyme A Mouse, anti-Human, R718, Clone: CB9, BD Horizon™

Catalog No. BDB566976 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB566976 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB566976 Supplier BD Biosciences Supplier No. 566976
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The CB9 monoclonal antibody specifically recognizes Granzyme A (GrA, GzmA or Gzm A) which is also known as CTL tryptase, Cytotoxic T-lymphocyte-associated serine esterase-3 (CTLA3), Cytotoxic T-lymphocyte proteinase 1, Fragmentin-1, or Hanukah factor serine protease (HFSP). Granzyme A belongs to a family of effector proteases that include five different granzymes that play key roles in innate and adaptive immune responses to pathogens and transformed cells: Granzymes A, B, H, K and M. Granzyme A is a ∼28 kDa neutral serine protease that is encoded by GZMA. It is expressed in the cytotoxic granules of natural killer (NK) cells, NKT cells, cytotoxic αâ T cells and ãä T cells. Granzyme A participates in innate and adaptive immune responses. After gaining access into target cells, Granzyme A cleaves various peptide substrates after lysine or arginine residues and can induce caspase‐independent cell death reflected in single-stranded DNA damage as well as rapid loss of cell membrane integrity and mitochondrial transmembrane potential.

TRUSTED_SUSTAINABILITY

Specifications

Antigen Granzyme A
Applications Flow Cytometry
Classification Monoclonal
Clone CB9
Conjugate R718
Host Species Mouse
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Refrigerate Upon Arrival
Form Liquid
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.