Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ Lab Vision™ Granzyme B Ab-1, Mouse Monoclonal Antibody

Catalog No. MS1157RQ Shop All Epredia Products
Change view
Click to view available options
Quantity:
100 μL
500 μL
1 mL
7 mL
12 mL
Form:
Concentrated
Prediluted
Ready-to-Use
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Form
MS1157RQ 12 mL Ready-to-Use
MS1157S1 500 μL Concentrated
MS1157S 1 mL Concentrated
MS1157S0 100 μL Concentrated
MS1157R1 1 mL Prediluted
MS1157R7 7 mL Prediluted
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. MS1157RQ Supplier Epredia™ Supplier No. MS1157RQ

Ensure accurate, reproducible results in immunohistochemistry experiments with Epredia™ Granzyme B Ab-1, Mouse Monoclonal Antibody.

Granzymes are neutral serine proteases, which are stored in specialized lytic granules of cytotoxic T lymphocytes (CTLs) and in natural killer (NK) cells. A number of granzymes (A to G) have been isolated and cloned from mouse CTLs and NK cells, however in man, fewer have been cloned and identified. One of these, human granzyme B, is involved in target cell apoptosis during lymphocyte-mediated cytotoxicity. Exocytosis of granzyme-containing granules in the cytoplasm of the target cell leads to the induction of DNA fragmentation and apoptosis.

Host Species: Mouse

Clone: GZB01

Isotype: IgG2a

Species Reactivity: Human. Others not known.

Epitope: N-terminal

Immunogen: Recombinant protein encoding the N-terminus of the mature granzyme B

Molecular Weight: 32 kDa

Positive Control: Hodgkin's lymphoma, Tonsil

Cellular Localization: Granular cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin only)

Specifications

Antigen Granzyme B Ab-1
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone GZB01
Conjugate Unconjugated
Host Species Mouse
Immunogen Recombinant protein encoding the N-terminus of the mature granzyme B
Quantity 12 mL
Regulatory Status IVD
Research Discipline Apoptosis
Primary or Secondary Primary
Form Ready-to-Use
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.