Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Granzyme B Antibody (B18.1), PE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP197525PE
Description
Granzyme B Monoclonal specifically detects Granzyme B in Human samples. It is validated for Flow Cytometry.Specifications
Granzyme B | |
Monoclonal | |
PE | |
C11, CCPI, CGL1, CGL-1, CSPB, CSP-B, CTLA1, CTSGL1, granzyme B, GranzymeB, GZMB, HLP, SECT | |
Mouse | |
Protein G purified | |
RUO | |
3002 | |
Store at 4C in the dark. | |
IgG1 |
Flow Cytometry | |
B18.1 | |
Flow Cytometry | |
GZMB | |
Recombinant human granzyme B (aa 7-227). | |
0.1 mL | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction