Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Granzyme B Antibody - BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody
$206.50 - $499.50
Specifications
Antigen | Granzyme B |
---|---|
Concentration | 1.0 mg/mL |
Dilution | Western Blot 0.5 - 1.0 ug/ml, Flow Cytometry 1-5 ug/ml, Immunohistochemistry 1:500, Immunohistochemistry-Paraffin 1:500 |
Applications | Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP259678SS
![]() |
Novus Biologicals
NBP259678SS |
0.025 mg |
Each for $206.50
|
|
|||||
NBP259678
![]() |
Novus Biologicals
NBP259678 |
0.1 mg |
Each for $499.50
|
|
|||||
Description
Granzyme B Polyclonal specifically detects Granzyme B in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
Granzyme B | |
Western Blot 0.5 - 1.0 ug/ml, Flow Cytometry 1-5 ug/ml, Immunohistochemistry 1:500, Immunohistochemistry-Paraffin 1:500 | |
Polyclonal | |
Rabbit | |
Apoptosis | |
PBS with 0.02% Sodium Azide | |
3002 | |
Partial recombinant protein made to the human Granzyme B protein (amino acids 19-247) [UniProt P10144] | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
1.0 mg/mL | |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
RUO | |
Human | |
C11, CCPI, CGL1, CGL-1, CSPB, CSP-B, CTLA1, CTSGL1, granzyme B, GranzymeB, GZMB, HLP, SECT | |
GZMB | |
IgG | |
Affinity Purified |
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title