Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Granzyme B Antibody (GB11), Novus Biologicals™
SDP

Catalog No. NB017639 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.05 mg
0.1 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB017639 0.025 mg
NB100647745 0.05 mg
NB10064774 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. NB017639 Supplier Novus Biologicals Supplier No. NB100647740.025MG
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Granzyme B Monoclonal specifically detects Granzyme B in Human, Primate, Monkey samples. It is validated for Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Frozen.

Specifications

Antigen Granzyme B
Applications Flow Cytometry, Immunofluorescence, Immunocytochemistry, Immunohistochemistry, Immunoprecipitation, ELISA
Classification Monoclonal
Clone GB11
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Flow Cytometry 1:10-1:100, ELISA 2 ug/ml-5 ug/ml, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500
Formulation Tris buffered saline with 0.09% Sodium Azide
Gene Accession No. P10144
Gene Alias C11, CCPI, CGL1, CGL-1, CSPB, CSP-B, CTLA1, CTSGL1, granzyme B, GZMB, HLP, SECT
Gene Symbols GZMB
Host Species Mouse
Immunogen Purified human Granzyme B (GZMB)
Purification Method Protein A purified
Quantity 0.025 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3002
Test Specificity NB100-64774 recognizes the serine protease Granzyme B (GZMB)
Target Species Primate, Monkey, Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.