Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Granzyme B Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody has been used in 5 publications

$258.08 - $862.71

Specifications

Antigen Granzyme B
Dilution Immunohistochemistry 1:50-1:100, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunohistochemistry-Paraffin 1:50-1:100, Immunohistochemistry-Frozen 1:10-1:500
Applications Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB100684SS
SDP
View Documents
Novus Biologicals
NB100684SS
0.1 mL
Each for $258.08
Only null left
Add to Cart
 
NB100684
SDP
View Documents
Novus Biologicals
NB100684
0.5 mL
Each for $862.71
Only null left
Add to Cart
 
Description

Description

Granzyme B Polyclonal specifically detects Granzyme B in Human, Mouse, Rat samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
Specifications

Specifications

Granzyme B
Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
Unconjugated
RUO
PBS (pH 7.4), 0.2% BSA, Tween-20 with 0.05% Sodium Azide
3002
Synthetic peptide: EIIGGHEAKPHSRPYMAYL, corresponding to amino acids 20-38 of Human Granzyme B (N-terminus).
Primary
Store at 4C. Do not freeze.
Immunohistochemistry 1:50-1:100, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunohistochemistry-Paraffin 1:50-1:100, Immunohistochemistry-Frozen 1:10-1:500
Polyclonal
Rabbit
Human, Mouse, Rat
C11, CCPI, CGL1, CGL-1, CSPB, CSP-B, CTLA1, CTSGL1, granzyme B, GranzymeB, GZMB, HLP, SECT
GZMB
IgG
Affinity Purified
This reacts with a 32 kDa protein.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.