Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Granzyme B Mouse Anti-Human, RY610, Clone: GB11, BD Horizon™
Mouse Monoclonal Antibody
$228.00 - $406.00
Specifications
Antigen | Granzyme B |
---|---|
Clone | GB11 |
Applications | Flow Cytometry (Intracellular Staining) |
Classification | Monoclonal |
Conjugate | RY610 |
Description
Granzyme B Monoclonal antibody specifically detects Granzyme B in Human (QC Testing) samples. It is validated for Flow Cytometry (Intracellular Staining)Specifications
Granzyme B | |
Flow Cytometry (Intracellular Staining) | |
RY610 | |
Mouse | |
Human | |
GZMB; Granzyme-2; CCPI; CGL1; CSPB; CTLA1; CTSGL1; GRB; HLP; SECT | |
Human Granzyme B | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
GB11 | |
Monoclonal | |
Purified | |
RUO | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
3002 | |
IgG1 κ | |
The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title