Learn More
Granzyme B Recombinant Rabbit Monoclonal Antibody (BLR022E), Bethyl Laboratories
Rabbit Recombit Monoclonal Antibody
$157.08 - $595.98
Specifications
Antigen | Granzyme B |
---|---|
Clone | BLR022E |
Applications | Immunocytochemistry, Immunohistochemistry, Western Blot |
Classification | Recombinant Monoclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-157-1298
![]() |
Bethyl Laboratories, Inc
A700022T |
10 μL |
Each for $157.08
|
|
|||||
50-157-1297
![]() |
Bethyl Laboratories, Inc
A700022 |
100 μL |
Each for $595.98
|
|
|||||
Description
The recommended shelf life for this product is 1 year from date of receipt. Application Note: For ICC, epitope retrieval with citrate buffer pH 6.0 is recommended for FFPE cell sections. For IHC, epitope retrieval with citrate buffer pH 6.0 is recommende
Granzyme B is a member of the granzyme serine protease family, and is found in the granules of cytotoxic T cells and NK cells. Granzyme B has been described as CGL1 (cathepsin G-like-1), a serine protease expressed only in cytotoxic T-lymphocytes after cell activation, and CTLA-1 (cytotoxic T lymphocyte-associated serine esterase 1) based on identification of mRNA in various cytotoxic T cells, but not observed in non-cytotoxic lymphoid cells. Granzyme B is crucial for the rapid induction of target cell death by apoptosis, induced by interaction with cytotoxic T cells. The receptor involved in this process has been identified as mannose 6-phosphate receptor which functions as a death receptor for Granzyme B during cytotoxic T cell-induced apoptosis. Granzyme B enters target cells to cleave caspase-3 and initiate the caspase cascade leading to DNA fragmentation and apoptosis. Granzyme B can also act through a mitochondrial apoptosis pathway by cleaving the Bid protein. Granzymes are neutral serine proteases, which are stored in specialized lytic granules of cytotoxic T lymphocytes (CTLs) and in natural killer (NK) cells. A number of granzymes (A to G) have been isolated and cloned from mouse CTLs and NK cells, however in man, fewer have been cloned and identified.Specifications
Granzyme B | |
Immunocytochemistry, Immunohistochemistry, Western Blot | |
Unconjugated | |
Rabbit | |
Human | |
P10144 | |
3002 | |
Recombit protein representing Human Granzyme B | |
Primary | |
4° C | |
GZMB |
BLR022E | |
Recombinant Monoclonal | |
Liquid | |
RUO | |
BBS with 0.1% BSA and 0.09% sodium azide; pH 8.2 | |
GrB | |
GZMB | |
IgG | |
Protein A | |
Antibody |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.